Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mec-7
Entrez Gene ID
181036
Gene Name
Tubulin beta-1 chain
Gene Aliases
CELE_ZK154.3
Chromosome Location
Chr.X: 7774856 - 7776689 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076912.5 NP_509313.1 2 - 3 307 57

View Full Product Details