Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sfxn-2
Entrez Gene ID
181054
Gene Name
Sidoreflexin
Gene Aliases
CELE_C14F5.4
Chromosome Location
Chr.X: 7956188 - 7958303 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076940.8 NP_509341.2 5 - 6 825 87

View Full Product Details