Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vem-1
Entrez Gene ID
181066
Gene Name
Protein vem-1
Gene Aliases
CELE_K07E3.8
Chromosome Location
Chr.X: 8087553 - 8089657 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029599.10 NP_001024770.1 3 - 4 686 76

View Full Product Details