Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cdd-1
Entrez Gene ID
183546
Gene Name
Cytidine deaminase
Gene Aliases
CELE_C47D2.2
Chromosome Location
Chr.X: 8208748 - 8211149 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076983.5 NP_509384.1 3 - 4 605 67

View Full Product Details