Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kqt-2
Entrez Gene ID
181078
Gene Name
Ion_trans domain-containing protein
Gene Aliases
CELE_M60.5
Chromosome Location
Chr.X: 8243131 - 8246369 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076991.7 NP_509392.3 13 - 14 1918 67

View Full Product Details