Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyn-8
Entrez Gene ID
181136
Gene Name
Peptidyl-prolyl cis-trans isomerase 8
Gene Aliases
CELE_D1009.2
Chromosome Location
Chr.X: 8911293 - 8914261 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077106.2 NP_509507.1 2 - 3 675 81

View Full Product Details