Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gly-13
Entrez Gene ID
181160
Gene Name
Putative alpha-1,3-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase
Gene Aliases
CELE_B0416.6
Chromosome Location
Chr.X: 9293657 - 9296622 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077165.7 NP_509566.1 11 - 12 1169 74
GenBank mRNA AF082010.1 - 11 - 12 1169 74

View Full Product Details