Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
apa-2
Entrez Gene ID
181163
Gene Name
AP-2 complex subunit alpha
Gene Aliases
CELE_T20B5.1
Chromosome Location
Chr.X: 9338400 - 9343361 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383622.1 7 - 8 2483 109

View Full Product Details