Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cut-5
Entrez Gene ID
187677
Gene Name
protein-coding
Gene Aliases
CELE_R07E3.3
Chromosome Location
Chr.X: 10333858 - 10338200 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270170.4 NP_001257099.1 8 - 9 923 109
NM_001270171.3 NP_001257100.1 7 - 8 960 109

View Full Product Details