Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mig-15
Entrez Gene ID
181248
Gene Name
protein-coding
Gene Aliases
CELE_ZC504.4
Chromosome Location
Chr.X: 10421653 - 10428657 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029800.3 NP_001024971.1 9 - 10 2027 77
NM_001029801.5 NP_001024972.1 9 - 10 2012 77
NM_001029802.5 NP_001024973.1 9 - 10 1951 77
NM_001029803.6 NP_001024974.1 10 - 11 1989 77
GenBank mRNA AF087131.1 - 9 - 10 2049 77

View Full Product Details