Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lpr-4
Entrez Gene ID
181298
Gene Name
LiPocalin-Related protein
Gene Aliases
CELE_W04G3.3
Chromosome Location
Chr.X: 11065126 - 11066811 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077451.7 NP_509852.2 4 - 5 883 77

View Full Product Details