Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sulp-7
Entrez Gene ID
189195
Gene Name
protein-coding
Gene Aliases
CELE_W04G3.6
Chromosome Location
Chr.X: 11080678 - 11087474 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038479.2 NP_001033568.1 2 - 3 131 127
NM_001038480.2 NP_001033569.1 2 - 3 191 127
NM_001038481.4 NP_001033570.1 4 - 5 463 127
NM_001038482.4 NP_001033571.1 4 - 5 463 127
GenBank mRNA AY887913.1 - 2 - 3 179 127
AY887914.1 - 2 - 3 236 127
AY887915.1 - 4 - 5 485 127
AY887916.1 - 4 - 5 485 127

View Full Product Details