Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sulp-7
Entrez Gene ID
189195
Gene Name
protein-coding
Gene Aliases
CELE_W04G3.6
Chromosome Location
Chr.X: 11080678 - 11087474 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038479.2 NP_001033568.1 5 - 6 483 113
NM_001038480.2 NP_001033569.1 5 - 6 543 113
NM_001038482.4 NP_001033571.1 7 - 8 815 113
GenBank mRNA AY887913.1 - 5 - 6 531 113
AY887914.1 - 5 - 6 588 113
AY887916.1 - 7 - 8 837 113

View Full Product Details