Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gnrr-6
Entrez Gene ID
184413
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F13D2.2
Chromosome Location
Chr.X: 11178899 - 11181274 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077464.4 NP_509865.1 4 - 5 654 129

View Full Product Details