Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cutl-11
Entrez Gene ID
188066
Gene Name
ZP domain-containing protein
Gene Aliases
CELE_T04F8.4
Chromosome Location
Chr.X: 11648188 - 11650851 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001375143.2 NP_001361820.1 5 - 5 441 76

View Full Product Details