Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-2
Entrez Gene ID
181400
Gene Name
protein-coding
Gene Aliases
CELE_F17E5.1
Chromosome Location
Chr.X: 12399517 - 12414041 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029416.4 NP_001024587.1 18 - 19 2104 69
NM_001029417.4 NP_001024588.1 8 - 9 1065 69
GenBank mRNA X92564.1 - - 2088 69
X92565.1 - - 1065 69

View Full Product Details