Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-26
Entrez Gene ID
188212
Gene Name
Nuclear Hormone Receptor family
Gene Aliases
CELE_T07C5.5
Chromosome Location
Chr.X: 12695229 - 12700465 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077718.5 NP_510119.1 2 - 3 2073 104

View Full Product Details