Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- bcat-1
- Entrez Gene ID
-
181423
- Gene Name
- Branched-chain-amino-acid aminotransferase, cytosolic
- Gene Aliases
-
CELE_K02A4.1
- Chromosome Location
-
Chr.X: 12877480 - 12880328 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077743.5 |
NP_510144.1 |
2 - 3 |
338 |
96 |
View Full Product Details