Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pqn-18
Entrez Gene ID
181425
Gene Name
protein-coding
Gene Aliases
CELE_C34E7.1
Chromosome Location
Chr.X: 12907825 - 12920260 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381127.1 NP_001366740.1 2 - 3 594 98

View Full Product Details