Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tre-4
Entrez Gene ID
184505
Gene Name
Trehalase
Gene Aliases
CELE_F15A2.2
Chromosome Location
Chr.X: 13472155 - 13475931 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077848.8 NP_510249.2 7 - 8 1134 151
GenBank mRNA AJ512338.1 - 7 - 8 1134 151

View Full Product Details