Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-51
Entrez Gene ID
184506
Gene Name
Serpentine receptor class delta-51
Gene Aliases
CELE_F15A2.3
Chromosome Location
Chr.X: 13482649 - 13484913 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077851.2 NP_510252.1 1 - 2 344 141

View Full Product Details