Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
odr-1
Entrez Gene ID
181479
Gene Name
Guanylate cyclase
Gene Aliases
CELE_R01E6.1
Chromosome Location
Chr.X: 13544729 - 13550990 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001375164.2 NP_001362115.1 21 - 21 2958 102

View Full Product Details