Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-23
Entrez Gene ID
192071
Gene Name
protein-coding
Gene Aliases
CELE_F19D8.1
Chromosome Location
Chr.X: 13817500 - 13823582 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270300.3 NP_001257229.1 10 - 11 1186 126
NM_001270301.4 NP_001257230.1 9 - 10 1090 126

View Full Product Details