Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-37
Entrez Gene ID
186978
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_K04C1.1
Chromosome Location
Chr.X: 14233944 - 14235032 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373569.1 NP_001359773.1 3 - 4 831 71

View Full Product Details