Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
thn-2
Entrez Gene ID
178187
Gene Name
THaumatiN family
Gene Aliases
CELE_F28D1.5
Chromosome Location
Chr.IV: 12383170 - 12383974 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069961.6 NP_502362.1 1 - 2 259 177

View Full Product Details