Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-34
Entrez Gene ID
177188
Gene Name
Cuticle collagen 34
Gene Aliases
CELE_F36A4.10
Chromosome Location
Chr.IV: 4243545 - 4244610 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068119.6 NP_500520.1 2 - 3 749 142
GenBank mRNA AF410845.1 - 2 - 3 749 142

View Full Product Details