Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxb-98
Entrez Gene ID
182660
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_C16C4.6
Chromosome Location
Chr.II: 1862634 - 1863782 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061699.1 NP_494100.1 1 - 2 250 136

View Full Product Details