Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-13A4
Entrez Gene ID
188355
Gene Name
Putative cytochrome P450 CYP13A4
Gene Aliases
CELE_T10B9.1
Chromosome Location
Chr.II: 9795756 - 9797740 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063710.3 NP_496111.1 1 - 2 181 112

View Full Product Details