Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bath-29
Entrez Gene ID
184454
Gene Name
protein-coding
Gene Aliases
CELE_F14D2.4
Chromosome Location
Chr.II: 3348560 - 3358983 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026914.4 NP_001022085.1 2 - 2 170 88
NM_001026915.3 NP_001022086.1 1 - 1 46 88

View Full Product Details