Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-25
Entrez Gene ID
3565817
Gene Name
C-type lectin domain-containing protein
Gene Aliases
CELE_T20B3.15
Chromosome Location
Chr.V: 16843241 - 16844851 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028978.3 NP_001024149.1 5 - 6 769 112

View Full Product Details