Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- math-37
- Entrez Gene ID
-
187883
- Gene Name
- MATH (meprin-associated Traf homology) domain containing
- Gene Aliases
-
CELE_R52.9
- Chromosome Location
-
Chr.II: 2116884 - 2122191 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061787.5 |
NP_494188.4 |
1 - 2 |
116 |
137 |
View Full Product Details