Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-54
Entrez Gene ID
189275
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_W07A12.6
Chromosome Location
Chr.II: 9158108 - 9159984 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063552.5 NP_495953.3 1 - 2 129 85

View Full Product Details