Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
exc-4
Entrez Gene ID
173314
Gene Name
Chloride intracellular channel exc-4
Gene Aliases
CELE_Y105E8A.22
Chromosome Location
Chr.I: 14511431 - 14518719 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170954.3 NP_740950.2 2 - 3 514 110
GenBank mRNA AY308063.1 - 2 - 3 510 110

View Full Product Details