Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-38
Entrez Gene ID
3565113
Gene Name
protein-coding
Gene Aliases
CELE_C35D6.9
Chromosome Location
Chr.IV: 16335337 - 16336503 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047456.1 NP_001040921.1 1 - 2 147 167

View Full Product Details