Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpd-3
Entrez Gene ID
180601
Gene Name
Glyceraldehyde-3-phosphate dehydrogenase 3
Gene Aliases
CELE_K10B3.7
Chromosome Location
Chr.X: 3112083 - 3114742 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076133.8 NP_508534.3 3 - 4 1568 80

View Full Product Details