Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- abu-7
- Entrez Gene ID
-
182139
- Gene Name
- Activated in Blocked Unfolded protein response
- Gene Aliases
-
CELE_C03A7.8
- Chromosome Location
-
Chr.V: 5163270 - 5164782 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071983.2 |
NP_504384.1 |
2 - 2 |
670 |
142 |
- Gene Symbol
- abu-6
- Entrez Gene ID
-
178903
- Gene Name
- Activated in Blocked Unfolded protein response
- Gene Aliases
-
CELE_C03A7.7
- Chromosome Location
-
Chr.V: 5160678 - 5164726 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071982.5 |
NP_504383.1 |
2 - 2 |
614 |
142 |
View Full Product Details