Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MED26
Entrez Gene ID
484847
Gene Name
mediator complex subunit 26
Gene Aliases
CRSP7
Chromosome Location
Chr.20: 45923034 - 45977118 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_541963.6 XP_541963.3 1 - 2 262 51

View Full Product Details