Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LEP
Entrez Gene ID
403616
Gene Name
leptin
Gene Aliases
OB
Chromosome Location
Chr.14: 8115396 - 8130946 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003070.1 NP_001003070.1 1 - 2 174 67
XM_005628340.3 XP_005628397.2 2 - 3 265 67
XM_005628341.2 XP_005628398.1 4 - 5 400 67
XM_022427124.1 XP_022282832.1 3 - 4 277 67
GenBank mRNA AB020986.1 - 1 - 2 174 67
U95642.1 - 1 - 2 80 67

View Full Product Details