Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AICDA
Entrez Gene ID
442983
Gene Name
activation induced cytidine deaminase
Gene Aliases
AID
Chromosome Location
Chr.27: 37064107 - 37073791 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003380.1 NP_001003380.1 2 - 3 228 76
XM_022410855.1 XP_022266563.1 1 - 2 282 76
GenBank mRNA AB122019.1 - 2 - 3 228 76

View Full Product Details