Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CALCB
Entrez Gene ID
403415
Gene Name
calcitonin-related polypeptide beta
Gene Aliases
CRSP-1, CRSP1
Chromosome Location
Chr.21: 37658360 - 37661361 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002948.2 NP_001002948.1 1 - 2 96 118
XM_022407411.1 XP_022263119.1 1 - 2 297 118
GenBank mRNA AB125102.1 - 1 - 2 97 118

View Full Product Details