Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLUL1
Entrez Gene ID
403747
Gene Name
clusterin like 1
Gene Aliases
-
Chromosome Location
Chr.7: 67431919 - 67483312 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003135.2 NP_001003135.2 7 - 8 1302 89
XM_014115499.2 XP_013970974.2 8 - 9 3584 89
XM_022420728.1 XP_022276436.1 8 - 9 3584 89
XM_022420729.1 XP_022276437.1 8 - 9 1568 89
XM_022420730.1 XP_022276438.1 6 - 7 2398 89
GenBank mRNA AF147784.2 - - 803 89
AF241221.1 - 7 - 8 1302 89

View Full Product Details