Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYL2
Entrez Gene ID
403614
Gene Name
myosin light chain 2
Gene Aliases
-
Chromosome Location
Chr.26: 8624147 - 8632166 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003069.1 NP_001003069.1 - 32 62
GenBank mRNA AF440218.1 - - 32 62

View Full Product Details