Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELSPBP1
Entrez Gene ID
399530
Gene Name
epididymal sperm binding protein 1
Gene Aliases
E12, E12A
Chromosome Location
Chr.1: 108159609 - 108187224 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002931.1 NP_001002931.1 6 - 7 659 71
XM_005615714.3 XP_005615771.1 5 - 6 688 71
XM_005615715.3 XP_005615772.1 4 - 5 552 71
XM_022403243.1 XP_022258951.1 6 - 7 772 71
XM_022403836.1 XP_022259544.1 5 - 6 704 71
GenBank mRNA AJ278477.1 - 6 - 7 659 71

View Full Product Details