Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM47
Entrez Gene ID
403423
Gene Name
RNA binding motif protein 47
Gene Aliases
-
Chromosome Location
Chr.3: 71985725 - 72135271 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002955.1 NP_001002955.1 3 - 4 1533 65
XM_005618245.3 XP_005618302.1 3 - 4 1540 65
XM_005618246.3 XP_005618303.1 3 - 4 1537 65
GenBank mRNA AJ388555.1 - 3 - 4 1533 65

View Full Product Details