Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTH
Entrez Gene ID
403986
Gene Name
parathyroid hormone
Gene Aliases
-
Chromosome Location
Chr.21: 36417062 - 36419967 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003302.1 NP_001003302.1 2 - 3 168 148
GenBank mRNA U15662.1 - 2 - 3 168 148

View Full Product Details