Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HRH3
Entrez Gene ID
477282
Gene Name
histamine receptor H3
Gene Aliases
-
Chromosome Location
Chr.24: 46239883 - 46243123 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005635258.2 XP_005635315.2 1 - 2 205 97
GenBank mRNA AH012621.2 - 1 - 2 205 97

View Full Product Details