Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TYR
Entrez Gene ID
403405
Gene Name
tyrosinase
Gene Aliases
-
Chromosome Location
Chr.21: 10799940 - 10894187 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002941.1 NP_001002941.1 4 - 5 1402 104
GenBank mRNA AY336053.1 - 4 - 5 1402 104

View Full Product Details