Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAS1R2
Entrez Gene ID
478213
Gene Name
taste 1 receptor member 2
Gene Aliases
-
Chromosome Location
Chr.2: 79719625 - 79738500 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031819.1 NP_001026989.1 5 - 6 1593 77
GenBank mRNA AY916758.1 - 5 - 6 1593 77

View Full Product Details