Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPCS2
Entrez Gene ID
404016
Gene Name
signal peptidase complex subunit 2
Gene Aliases
SPC25
Chromosome Location
Chr.21: 23485367 - 23510249 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003324.2 NP_001003324.1 2 - 3 221 99
GenBank mRNA U12687.1 - 2 - 3 222 99

View Full Product Details