Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSPB1
Entrez Gene ID
403979
Gene Name
heat shock protein family B (small) member 1
Gene Aliases
HSP27
Chromosome Location
Chr.6: 7487164 - 7488576 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003295.2 NP_001003295.2 - 543 82
GenBank mRNA KJ947842.1 - - 436 82
U19368.1 - - 476 82

View Full Product Details